missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR5AK2 Polyclonal Antibody

Description
OR5AK2 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
Specifications
| Antigen | OR5AK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OR5AK2 |
| Gene Accession No. | Q8NH90 |
| Gene Alias | Olfactory receptor 5AK2; olfactory receptor family 5 subfamily AK member 2; olfactory receptor, family 5, subfamily AK, member 2; OR5AK2 |
| Gene Symbols | OR5AK2 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?