missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR5AC2 Polyclonal Antibody

Product Code. 15683545
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15683545 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15683545 Supplier Invitrogen™ Supplier No. PA549254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR5AC2 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR5AC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR5AC2
Gene Accession No. Q9NZP5
Gene Alias HSA1; Olfactory receptor 5AC2; olfactory receptor family 5 subfamily AC member 2; olfactory receptor, family 5, subfamily AC, member 2; OR5AC2
Gene Symbols OR5AC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 246-274 amino acids from the C-terminal region of human OR5AC2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.