missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OR52N5 Polyclonal specifically detects OR52N5 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | OR52N5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | olfactory receptor 52N5, Olfactory receptor OR11-62, olfactory receptor, family 52, subfamily N, member 5, OR11-62, OR52N5Q |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR52N5 (NP_001001922). Peptide sequence GHTIPPSLHIIVANLYLLLPPTLNPIVYGVKTKQIRKSVIKFFQGDKGAG |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?