missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR52L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548764
This item is not returnable.
View return policy
Description
OR52L1 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
| OR52L1 | |
| Polyclonal | |
| Unconjugated | |
| OR52L1 | |
| Olfactory receptor 52L1; olfactory receptor family 52 subfamily L member 1; olfactory receptor OR11-50; olfactory receptor, family 52, subfamily L, member 1; OR11-50; OR52L1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 338751 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NGH7 | |
| OR52L1 | |
| KLH conjugated synthetic peptide between 300-329 amino acids from the C-terminal region of human OR52L1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction