missing translation for 'onlineSavingsMsg'
Learn More

OR52I2 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18358573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358573 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358573 Supplier Novus Biologicals Supplier No. NBP306488UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR52I2 Polyclonal antibody specifically detects OR52I2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR52I2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias olfactory receptor 52I2, olfactory receptor, family 52, subfamily I, member 2, OR11-12
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OR52I2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143502
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.