missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR52B6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-09866-100UL
This item is not returnable.
View return policy
Description
OR52B6 Polyclonal specifically detects OR52B6 in Human samples. It is validated for Western Blot.
Specifications
| OR52B6 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| olfactory receptor 52B6, Olfactory receptor OR11-47, olfactory receptor, family 52, subfamily B, member 6, OR11-47 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR52B6 (NP_001005162). Peptide sequence VWYGLAAALLSTGLDIMLITVSYIHILQAVFRLLSQDARSKALSTCGSHI | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 340980 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction