missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR51M1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100966
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR51M1.
Odorant receptor.
Specifications
| OR51M1 | |
| Polyclonal | |
| Unconjugated | |
| OR51M1 | |
| HOR5'Beta7; Odorant receptor HOR5'beta7; olfactory receptor 51M1; olfactory receptor family 51 subfamily M member 1; olfactory receptor OR11-40; olfactory receptor, family 51, subfamily M, member 1; OR11-40; OR51M1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 390059 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H341 | |
| OR51M1 | |
| A synthesized peptide derived from human OR51M1(Accession Q9H341), corresponding to amino acid residues L257-K307. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction