missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR4K15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563166
This item is not returnable.
View return policy
Description
Immunogen sequence: KLKSRYLKPS QVSVVIRNVL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 80%.
Specifications
| OR4K15 | |
| Polyclonal | |
| Unconjugated | |
| OR4K15 | |
| Olfactory receptor 4K15; olfactory receptor family 4 subfamily K member 15; olfactory receptor OR14-20; olfactory receptor, family 4, subfamily K, member 15; OR14-20; OR4K15; OR4K15Q | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NH41 | |
| OR4K15 | |
| Recombinant protein corresponding to Human OR4K15. Recombinant protein control fragment (Product #RP-100765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction