missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2W5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569316
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. This olfactory receptor gene has a coding sequence that is comparable in length to other olfactory receptor genes, but it should be noted that a frameshift is present in the 3' coding region that disrupts the 7-transmembrane domain structure in the protein. It is unclear if the protein can function as an olfactory receptor or if an alternate function is served. For this reason, this gene has also been interpreted to be a pseudogene.
Specifications
| OR2W5 | |
| Polyclonal | |
| Unconjugated | |
| OR2W5 | |
| olfactory receptor 2W5; olfactory receptor family 2 subfamily W member 5 (gene/pseudogene); olfactory receptor, family 2, subfamily W, member 5 pseudogene; OR2W5; OR2W5P; OST722; Putative olfactory receptor 2W5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 441932 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6NFC9 | |
| OR2W5 | |
| synthetic peptide directed towards the middle region of human OR2W5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction