missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2V2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100990
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR2V2.
Olfactory receptors interact with odorant molecules in thenose, to initiate a neuronal response that triggers the perceptionof a smell. The olfactory receptor proteins are members of a largefamily of G-protein-coupled receptors (GPCR) arising from singlecoding-exon genes. Olfactory receptors share a 7-transmembranedomain structure with many neurotransmitter and hormone receptorsand are responsible for the recognition and G protein-mediatedtransduction of odorant signals. The olfactory receptor gene familyis the largest in the genome. The nomenclature assigned to theolfactory receptor genes and proteins for this organism isindependent of other organisms.
Specifications
| OR2V2 | |
| Polyclonal | |
| Unconjugated | |
| OR2V2 | |
| olfactory receptor 2V2; Olfactory receptor 2V3; olfactory receptor family 2 subfamily V member 2; olfactory receptor OR5-3; olfactory receptor, family 2, subfamily V, member 2; olfactory receptor, family 2, subfamily V, member 3; OR2V2; OR2V3; OST713 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285659 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96R30 | |
| OR2V2 | |
| A synthesized peptide derived from human OR2V2(Accession Q96R30), corresponding to amino acid residues A238-P288. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering