missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2M4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83314-0.1ML
This item is not returnable.
View return policy
Description
OR2M4 Polyclonal specifically detects OR2M4 in Human samples. It is validated for Western Blot.
Specifications
| OR2M4 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 26245 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| HSHTPCRX18, HTPCRX18Olfactory receptor TPCR100, novel 7 transmembrane receptor (rhodopsin family) protein, olfactory receptor 2M4, Olfactory receptor OR1-55, olfactory receptor, family 2, subfamily M, member 4, OST710OR1-55, TPCR100 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR2M4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction