missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2M4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100985
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR2M4.
Odorant receptor.
Specifications
| OR2M4 | |
| Polyclonal | |
| Unconjugated | |
| OR2M4 | |
| HSHTPCRX18; HTPCRX18; novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2M4; olfactory receptor family 2 subfamily M member 4; olfactory receptor OR1-55; Olfactory receptor TPCR100; olfactory receptor, family 2, subfamily M, member 4; OR1-55; OR2M4; OST710; TPCR100 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26245 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96R27 | |
| OR2M4 | |
| A synthesized peptide derived from human OR2M4(Accession Q96R27), corresponding to amino acid residues T240-Y290. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction