missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599949
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR2L2.
Odorant receptor.
Specifications
| OR2L2 | |
| Polyclonal | |
| Unconjugated | |
| OR2L2 | |
| HSHTPCRH07; HTPCRH07; Olfactory receptor 2L12; olfactory receptor 2L2; Olfactory receptor 2L4; olfactory receptor family 2 subfamily L member 2; olfactory receptor OR1-48 pseudogene; olfactory receptor, family 2, subfamily L, member 12; olfactory receptor, family 2, subfamily L, member 2; olfactory receptor, family 2, subfamily L, member 4 pseudogene; OR1-48; OR2L12; OR2L2; OR2L4P | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26246 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NH16 | |
| OR2L2 | |
| A synthesized peptide derived from human OR2L2(Accession Q8NH16), corresponding to amino acid residues E110-V160. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction