missing translation for 'onlineSavingsMsg'
Learn More

OR2H1 Antibody, Novus Biologicals™

Product Code. 18335870 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335870 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335870 Supplier Novus Biologicals Supplier No. NBP286738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR2H1 Polyclonal specifically detects OR2H1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2H1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias family 2, subfamily H, member 6, Hs6M1-16, Olfactory receptor 2H6, olfactory receptor, family 2, subfamily H, member 1,6M1-16, olfactory receptor, family 2, subfamily H, member 8, OLFR42A-9004-14, OR2H8, OR6-2HS6M1-16
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C region of human OR2H1
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26716
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.