missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2C3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97856-50ul
This item is not returnable.
View return policy
Description
OR2C3 Polyclonal antibody specifically detects OR2C3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OR2C3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| olfactory receptor 2C3, Olfactory receptor 2C4, Olfactory receptor 2C5, Olfactory receptor OR1-30, olfactory receptor, family 2, subfamily C, member 3, olfactory receptor, family 2, subfamily C, member 4, olfactory receptor, family 2, subfamily C, member 5 pseudogene, OR2C4, OR2C5P, OST742 | |
| Produced in rabbits immunized with E. coli-derived Human OR2C3 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 81472 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering