missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2B6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104304
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR2B6.
Odorant receptor.
Specifications
| OR2B6 | |
| Polyclonal | |
| Unconjugated | |
| OR2B6 | |
| dJ408B20.2; Hs6M1-32; olfactory receptor 2B1; Olfactory receptor 2B5; olfactory receptor 2B6; Olfactory receptor 5-40; olfactory receptor 6-31; olfactory receptor family 2 subfamily B member 6; olfactory receptor OR6-4; olfactory receptor, family 2, subfamily B, member 1 pseudogene; olfactory receptor, family 2, subfamily B, member 5; olfactory receptor, family 2, subfamily B, member 6 pseudogene; OR2B1; OR2B1P; OR2B5; OR2B6; OR2B6P; OR5-40; OR5-41; OR6-31 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26212 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P58173 | |
| OR2B6 | |
| A synthesized peptide derived from human OR2B6(Accession P58173), corresponding to amino acid residues H131-V181. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur