missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2AJ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102122
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR2AJ1.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR2AJ1 | |
| Polyclonal | |
| Unconjugated | |
| OR2AJ1 | |
| OR2AJ1 | |
| A synthesized peptide derived from human OR2AJ1(Accession Q8NGZ0), corresponding to amino acid residues F206-I256. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NGZ0 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 127608 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction