missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR2AG1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | OR2AG1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OR2AG1 Polyclonal specifically detects OR2AG1 in Human samples. It is validated for Western Blot.Specifications
| OR2AG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| HT3, hT3 olfactory receptor, olfactory receptor 2AG1, Olfactory receptor 2AG3, Olfactory receptor OR11-79, olfactory receptor, family 2, subfamily AG, member 1, olfactory receptor, family 2, subfamily AG, member 3, OR11-79, OR2AG3 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR2AG1 (NP_001004489). Peptide sequence LAAILASYTQILLTVLHMPSNEGRKKALVTCSSHLTVVGMFYGAATFMYV | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 144125 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title