missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116289
This item is not returnable.
View return policy
Description
OR2A5 Polyclonal Antibody for Western Blot
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR2A5 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96R48 | |
| OR2A5 | |
| KLH conjugated synthetic peptide between 246-272 amino acids from the C-terminal region of human OR2A5. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| OR2A5 | |
| Olfactory receptor 2A26; olfactory receptor 2A5; Olfactory receptor 2A8; olfactory receptor 7-138/7-141; olfactory receptor family 2 subfamily A member 5; olfactory receptor, family 2, subfamily A, member 11 pseudogene; olfactory receptor, family 2, subfamily A, member 26; olfactory receptor, family 2, subfamily A, member 5; olfactory receptor, family 2, subfamily A, member 8; OR2A11P; OR2A26; OR2A5; OR2A8; OR7-138; OR7-141 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 393046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction