missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2A42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116195
This item is not returnable.
View return policy
Description
OR2A42 Polyclonal Antibody for Western Blot
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR2A42 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NGT9 | |
| OR2A1 | |
| KLH conjugated synthetic peptide between 282-310 amino acids from the C-terminal region of human OR2A42. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| OR2A1 | |
| Olfactory receptor 2A1/2A42; olfactory receptor family 2 subfamily A member 1; olfactory receptor family 2 subfamily A member 42; olfactory receptor OR7-16; Olfactory receptor OR7-19; olfactory receptor, family 2, subfamily A, member 1; olfactory receptor, family 2, subfamily A, member 42; OR2A1; OR2A42 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 346528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction