missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2A25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571171
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 79%; Rat: 86%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR2A25 | |
| Polyclonal | |
| Unconjugated | |
| OR2A25 | |
| Olfactory receptor 2A25; olfactory receptor 2A27; olfactory receptor family 2 subfamily A member 25; olfactory receptor, family 2, subfamily A, member 24 pseudogene; olfactory receptor, family 2, subfamily A, member 25 pseudogene; olfactory receptor, family 2, subfamily A, member 27; OR2A24P; OR2A25; OR2A25P; OR2A27 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 392138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A4D2G3 | |
| OR2A25 | |
| synthetic peptide directed towards the C-terminal region of human OR2A25. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction