missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR1K1 Polyclonal Antibody

OR1K1 Polyclonal Antibody
Brand: Invitrogen PA571166
This item is not returnable.
View return policy
Description
OR1K1 Polyclonal Antibody for Western Blot
Specifications
| OR1K1 | |
| Polyclonal | |
| Unconjugated | |
| OR1K1 | |
| hg99; Olfactory receptor 1K1; olfactory receptor family 1 subfamily K member 1; olfactory receptor, family 1, subfamily K, member 1; OR1K1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 392392 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NGR3 | |
| OR1K1 | |
| synthetic peptide directed towards the C-terminal region of human OR1K1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction