missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR1A1 Recombinant Rabbit Monoclonal Antibody (K01_2K54)
Description
OR1A1 Recombinant Monoclonal Antibody for Western Blot
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
| Antigen | OR1A1 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2K54 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | OR1A1 |
| Gene Accession No. | Q9P1Q5 |
| Gene Alias | Olfactory receptor 17-7; olfactory receptor 1A1; olfactory receptor family 1 subfamily A member 1; olfactory receptor OR17-11; olfactory receptor, family 1, subfamily A, member 1; OR17-7; OR1A1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?