missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OR1A1 Polyclonal specifically detects OR1A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | OR1A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | olfactory receptor 1A1, Olfactory receptor OR17-11, olfactory receptor, family 1, subfamily A, member 1, OR17-7Olfactory receptor 17-7 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human OR1A1 (NP_055380.2). Peptide sequence RSCIWLIAGSWVIGNANALPHTLLTASLSFCGNQEVANFYCDITPLLKLS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?