missing translation for 'onlineSavingsMsg'
Learn More

OR14J1 Antibody, Novus Biologicals™

Product Code. 18492171 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18492171 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18492171 Supplier Novus Biologicals Supplier No. NBP19222325ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR14J1 Polyclonal specifically detects OR14J1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR14J1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias bA150A6.4, hs6M1-28, olfactory receptor 14J1, Olfactory receptor 5U1, Olfactory receptor OR6-25, olfactory receptor, family 14, subfamily J, member 1, olfactory receptor, family 5, subfamily U member 1, olfactory receptor, family 5, subfamily U, member 1, OR5U1, OR6-25
Gene Symbols OR14J1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:YSLRNDSMKAALRKMLSKEELPQR
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442191
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.