missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OR11A1 Polyclonal specifically detects OR11A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | OR11A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | dJ994E9.6, family 11, subfamily A, member 2, Hs6M1-18, olfactory receptor 11A1, Olfactory receptor 11A2, Olfactory receptor OR6-30, olfactory receptor, family 11, subfamily A, member 1, OR11A2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR11A1 (NP_039225). Peptide sequence YVAPSAVHSQLLSKVFSLLYTVVTPLFNPVIYTMRNKEVHQALRKILCIK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?