missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR11A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104307
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR11A1.
Odorant receptor.
Specifications
| OR11A1 | |
| Polyclonal | |
| Unconjugated | |
| OR11A1 | |
| 6M1-18; dJ994E9.6; hs6M1-18; Olfactory receptor 11A1; olfactory receptor 11A2; olfactory receptor family 11 subfamily A member 1; olfactory receptor OR6-30; olfactory receptor, family 11, subfamily A, member 1; olfactory receptor, family 11, subfamily A, member 2; OR11A1; OR11A2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26531 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9GZK7 | |
| OR11A1 | |
| A synthesized peptide derived from human OR11A1(Accession Q9GZK7), corresponding to amino acid residues V146-A196. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction