missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR10Q1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100971
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OR10Q1.
Odorant receptor.
Specifications
| OR10Q1 | |
| Polyclonal | |
| Unconjugated | |
| OR10Q1 | |
| olfactory receptor 10Q1; olfactory receptor family 10 subfamily Q member 1; olfactory receptor OR11-233; olfactory receptor, family 10, subfamily Q, member 1; OR10Q1; OR11-233 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 219960 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NGQ4 | |
| OR10Q1 | |
| A synthesized peptide derived from human OR10Q1(Accession Q8NGQ4), corresponding to amino acid residues L250-K300. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction