missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR10J5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534044

Product Code. 15342681

  • £441.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gibbon, Elephant, Horse (100%) Gorilla, Hamster, Dog (94%) Mouse, Rat, Panda (88%) Bovine, Pig (81%).

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OR10J5
Polyclonal
Unconjugated
OR10J5
MOR23; Olfactory receptor 10J5; olfactory receptor family 10 subfamily J member 5; olfactory receptor OR1-28; olfactory receptor, family 10, subfamily J, member 5; OR10J5; OR1-28
Rabbit
Antigen Affinity Chromatography
RUO
127385
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.81 mg/mL
PBS with 0.1% sodium azide
Q8NHC4
OR10J5
Synthetic 16 amino acid peptide from 2nd extracellular domain of human OR10J5.
50 μg
Primary
Human, Horse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.