missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10J5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS3159
This item is not returnable.
View return policy
Description
OR10J5 Polyclonal antibody specifically detects OR10J5 in Human, Equine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| OR10J5 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Equine | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| olfactory receptor 10J5, olfactory receptor, family 10, subfamily J, member 5 | |
| Synthetic 16 amino acid peptide from 2nd extracellular domain of human OR10J5. | |
| 0.05 mL | |
| Olfactory | |
| 127385 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction