missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | OR10C1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656706
|
Novus Biologicals
NBP3-06509-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686706
|
Novus Biologicals
NBP3-06509-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OR10C1 Polyclonal antibody specifically detects OR10C1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| OR10C1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| hs6M1-17, olfactory receptor 10C1, Olfactory receptor 10C2, olfactory receptor OR6-31, olfactory receptor, family 10, subfamily C, member 1, olfactory receptor, family 10, subfamily C, member 2, OR10C1P, OR10C2, OR6-31, seven transmembrane helix receptor | |
| Produced in rabbits immunized with E. coli-derived Human OR10C1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 442194 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title