missing translation for 'onlineSavingsMsg'
Learn More

OR10C1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18367642
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367642 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367642 Supplier Bio-Techne Supplier No. NBP306509DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR10C1 Polyclonal antibody specifically detects OR10C1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR10C1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias hs6M1-17, olfactory receptor 10C1, Olfactory receptor 10C2, olfactory receptor OR6-31, olfactory receptor, family 10, subfamily C, member 1, olfactory receptor, family 10, subfamily C, member 2, OR10C1P, OR10C2, OR6-31, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human OR10C1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442194
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.