missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR-486, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Potent and selective inhibitor of catechol-O-methyl-transferase.
Specifications
Specifications
CAS | 7659-29-2 |
Molecular Formula | C6H4N2O6 |
Quantity | 50 mg |
Synonym | 3,5-dinitrocatechol, 3,5-dinitro-1,2-benzenediol, 3,5-dinitropyrocatechol, unii-vk0va22gy2, pyrocatechol, 3,5-dinitro, 1,2-benzenediol, 3,5-dinitro, vk0va22gy2, 3,5-dinitro-benzene-1,2-diol, 3,5-dinitrocatechol or-486, dnc |
InChI Key | VDCDWNDTNSWDFJ-UHFFFAOYSA-N |
SMILES | C1=C(C=C(C(=C1[N+](=O)[O-])O)O)[N+](=O)[O-] |
IUPAC Name | 3,5-dinitrobenzene-1,2-diol |
Molecular Weight (g/mol) | 200.106 |
PubChem CID | 3870203 |
Formula Weight | 200.11 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'