missing translation for 'onlineSavingsMsg'
Learn More

Optovin, MedChemExpress

Product Code. 16495986
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495986 100 mg 100mg
16451557 50 mg 50mg
16435368 10 mg 10mg
16408087 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495986 Supplier Medchem Express Supplier No. HY12809
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Optovin is a reversible photoactivated TRPA1 ligand that enables light-mediated neuronal excitation. Optovin activates TRPA1 via structure-dependent photochemical reactions with redox-sensitive cysteine residues.

Optovin is a reversible photoactivated TRPA1 ligand that enables light-mediated neuronal excitation. Optovin activates TRPA1 via structure-dependent photochemical reactions with redox-sensitive cysteine residues.

Specifications

Chemical Name or Material Optovin
CAS 348575-88-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H13N3OS2
Quantity 100 mg
Solubility Information DMSO : ≥ 37 mg/mL (117.31 mM)
SMILES O=C1NC(S/C1=C\C2=C(C)N(C3=CC=CN=C3)C(C)=C2)=S
Molecular Weight (g/mol) 315.41
Formula Weight 315.41
Percent Purity 96.47%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.