Learn More
Specac™ Opto-Physics Infrared Polarizer ZnSe 18mm
Product Code. 15230398
Encompass_Preferred
Click to view available options
Type:
Opto-Physics Infrared Polarizer ZnSe 18mm
Product Code. 15230398
Brand: Specac™ GS57503
This is a 18mm Clear Aperture ZnSe opto-physics polariser manufactured at 2500 lines/mm to provide acceptable polarisation performance at an economical price.
Specifications
Type Opto-Physics Infrared Polarizer ZnSe 18mm