Learn More
Specac™ Opto-Physics Infrared Polarizer CaF2 34mm
Product Code. 15250398
Encompass_Preferred
Click to view available options
Type:
Opto-Physics Infrared Polarizer CaF2 34mm
Product Code. 15250398
Brand: Specac™ GS57505
This is a 34mm Clear Aperture CaF2 opto-physics polariser manufactured at 2500 lines/mm to provide acceptable polarisation performance at an economical price.
Specifications
Type Opto-Physics Infrared Polarizer CaF2 34mm