Learn More
Specac™ Opto-Physics Infrared Polarizer CaF2 18mm
Product Code. 15210398
Encompass_Preferred
Change view
Click to view available options
Type:
Opto-Physics Infrared Polarizer CaF2 18mm
Product Code. Type
15210398 Opto-Physics Infrared Polarizer CaF2 18mm
1 options
Product Code. 15210398 Supplier Specac™ Supplier No. GS57501
Only null left
Add to basket
Add to basket
This is a 18mm Clear Aperture CaF2 opto-physics polariser manufactured at 2500 lines/mm to provide acceptable polarisation performance at an economical price.
Specifications
Type Opto-Physics Infrared Polarizer CaF2 18mm