Learn More
Specac™ Opto-Physics Infrared Polarizer BaF2 34mm
Product Code. 15260398
Encompass_Preferred
Change view
Click to view available options
Type:
Opto-Physics Infrared Polarizer BaF2 34mm
Product Code. Type
15260398 Opto-Physics Infrared Polarizer BaF2 34mm
1 options
Product Code. 15260398 Supplier Specac™ Supplier No. GS57506
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is a 34mm Clear Aperture BaF2 opto-physics polariser manufactured at 2500 lines/mm to provide acceptable polarisation performance at an economical price.
Specifications
Type Opto-Physics Infrared Polarizer BaF2 34mm