Learn More
OPTN Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-10837-100UL
Description
The OPTN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPTN. This antibody recognizes human, mouse, rat antigen. The OPTN antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
| OPTN | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 10133 | |
| IgG |