missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ XhoI, Fisher BioReagents™

Product Code. 11914852
Click to view available options
Quantity:
3000 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11914852

Brand: Fisher BioReagents BP80101

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'....C^T C G A G...3'
3'....G A G C T^C...5'

Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2 , 100mM KCl and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 50 - 100%
  • Buffer 3: 50 - 100%
  • Buffer 4: 20 - 50%
  • Buffer 5: 100%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of DNA fragments can be ligated and re-cut after a 50-fold over-digestion with XhoI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of XhoI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Note:
  • Supercoiled plasmids may require up to 5-fold more XhoI for complete digestion than linear DNA.
  • Compatible Ends: BauI, Eco88I, SalI, SmoI
  • Methylation Effects:
  • Dam: Never overlaps - no effect
  • Dcm: Never overlaps - no effect
  • CpG: Completely overlaps - cleavage impaired
  • CTCGAG

(Cleavage impaired)
EcoKI: Never overlaps - no effect
EcoBI: Never overlaps - no effect

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Components 10X OPTIZYME™ Buffer 5
Incubator Temperature 37°C
pH 7.4
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Xhol
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Quantity 3000 U
Cut Site C.TCGAG
Product Type XhoI
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.