missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ T4 Polynucleotide Kinase, Fisher BioReagents™

Product Code. 12877142
Click to view available options
Quantity:
500 U
Unit Size:
500 units
This item is not returnable. View return policy

Product Code. 12877142

Brand: Fisher BioReagents BP80981

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

OPTIZYME™ T4 PNK catalyzes the transfer of γ-phosphate from ATP to the 5'-OH group of DNA, RNA, oligonucleotides and nucleoside 3'-monophosphates.

Applications and Features:

  • OPTIZYME™ T4 PNK labels 5'-termini of nucleic acids (for use as: probes for hybridization or transcript mapping, primers for PCR or DNA sequencing, and markers for gel-electrophoresis)
  • 5'-phosphorylation of oligonucleotides, PCR products, and other DNA or RNA prior to ligation
  • Phosphorylation of PCR primers
  • Removal of 3'-phosphate groups
  • Active in buffers for OPTIZYME restriction enzymes, T4 DNA Ligase and Reverse Transcriptases

Supplied With:

  • 10X OPTIZYME T4 PNK Buffer 1 (for forward reaction): 500mM Tris-HCl (pH7.6 at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine
  • 10X OPTIZYME T4 PNK Buffer 2 (for exchange reaction): 500mM imidazole-HCl (pH6.4 at 25°C), 180mM MgCl2, 50mM DTT, 1mM spermidine, 1mM ADP
  • 24% PEG 6000 Sol for T4 PNK: 24% (w/v) polyethylene glycol 6000
Source: E. Coli cells with a cloned pse T gene of bacteriophage T4
Quality Control:
  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested for labeling of 5'-termini of DNA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Color Colorless
pH 7.5
For Use With (Application) Probes for hybridization or transcript mapping, primers for PCR or DNA sequencing and markers for Gel electrophoresis, removal of 3'-phosphate groups
Storage Buffer 10X OPTIZYME™ T4 PNK Buffer 1 (for forward reaction): 500mM Tris HCl (7.6 pH at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine, 10X OPTIZYME™ T4 PNK Buffer 2 (for exchange reaction): 500mM imidazole-HCl
Content And Storage -20°C
Source E. coli cells with a cloned pse T gene of bacteriophage T4
Quantity 500 U
Product Type T4 Polynucleotide Kinase
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.