missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ T4 DNA Ligase, Fisher BioReagents™
Click to view available options
Quantity:
100 U
500 U
Unit Size:
100 units
500 units
Description
OPTIZYME™ T4 DNA Ligase catalyzes the formation of phosphodiester bonds between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.
Applications and Features:
- OPTIZYME™ T4 DNA Ligase is used to clone restriction enzyme generated DNA fragments
- Fast sticky end ligation (10min at room temperature)
- Cloning PCR products
- Ligation of double-stranded oligonucleotide linkers or adaptors to DNA
- Site-directed mutagenesis
- Amplified fragment length polymorphism
- Ligase-mediated RNA detection
- Nick repair in duplex DNA, RNA or DNA/RNA hybrids
- Active in buffers for OPTIZYME restriction enzymes
Supplied with:
- 10X OPTIZYME T4 DNA Ligase Buffer : 400mM Tris-HCl, 100mM MgCl2, 100mM DTT, 5mM ATP (pH7.8 at 25°C)
- 50% OPTIZYME PEG 4000 Solution: 50% (w/v) polyethylene glycol
Quality control:
- Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free
- Functionally tested for ligation of cohesive and blunt end DNA fragments
Specifications
Specifications
| Concentration | 5 U/μL |
| Color | Colorless |
| pH | 7.5 |
| For Use With (Application) | Used to clone restriction Enzyme generated DNA fragments, cloning PCR products, ligation of double-stranded Oligonucleotide linkers or adaptors to DNA, site-directed mutagenesis, amplified fragment length polymorphism, ligase-mediated RNA detection, nick |
| Storage Buffer | OPTIZYME™ T4 DNA Ligase Buffer 10X: Tris HCl 400mM, MgCl2 100mM, DTT 100mM, ATP 5mM (7.8 pH at 25°C), OPTIZYME™ PEG 4000 Solution 50%: Polyethylene Glycol 50% (w/v) |
| Content And Storage | -20°C |
| Source | E. coli cells with a cloned gene 30 of bacteriophage T4 |
| Quantity | 500 U |
| Product Type | T4 DNA Ligase |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction