missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ T4 DNA Ligase, Fisher BioReagents™

Product Code. 12807152
Click to view available options
Quantity:
100 U
500 U
Unit Size:
100 units
500 units
This item is not returnable. View return policy

Product Code. 12807152

Brand: Fisher BioReagents BP80991

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

OPTIZYME™ T4 DNA Ligase catalyzes the formation of phosphodiester bonds between juxtaposed 5'-phosphate and 3'-hydroxyl termini in duplex DNA or RNA.

Applications and Features:

  • OPTIZYME™ T4 DNA Ligase is used to clone restriction enzyme generated DNA fragments
  • Fast sticky end ligation (10min at room temperature)
  • Cloning PCR products
  • Ligation of double-stranded oligonucleotide linkers or adaptors to DNA
  • Site-directed mutagenesis
  • Amplified fragment length polymorphism
  • Ligase-mediated RNA detection
  • Nick repair in duplex DNA, RNA or DNA/RNA hybrids
  • Active in buffers for OPTIZYME restriction enzymes

Supplied with:

  • 10X OPTIZYME T4 DNA Ligase Buffer : 400mM Tris-HCl, 100mM MgCl2, 100mM DTT, 5mM ATP (pH7.8 at 25°C)
  • 50% OPTIZYME PEG 4000 Solution: 50% (w/v) polyethylene glycol
Source: E. Coli cells with a cloned gene 30 of bacteriophage T4
Quality control:
  • Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free
  • Functionally tested for ligation of cohesive and blunt end DNA fragments

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Color Colorless
pH 7.5
For Use With (Application) Used to clone restriction Enzyme generated DNA fragments, cloning PCR products, ligation of double-stranded Oligonucleotide linkers or adaptors to DNA, site-directed mutagenesis, amplified fragment length polymorphism, ligase-mediated RNA detection, nick
Storage Buffer OPTIZYME™ T4 DNA Ligase Buffer 10X: Tris HCl 400mM, MgCl2 100mM, DTT 100mM, ATP 5mM (7.8 pH at 25°C), OPTIZYME™ PEG 4000 Solution 50%: Polyethylene Glycol 50% (w/v)
Content And Storage -20°C
Source E. coli cells with a cloned gene 30 of bacteriophage T4
Quantity 100 U
Product Type T4 DNA Ligase
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.