missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ NheI, Fisher BioReagents™

Product Code. 11944782
Click to view available options
Quantity:
500 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11944782

Brand: Fisher BioReagents BP80191

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...G^C T A G C...3'
3'...C G A T C^G...5'

Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 100%
  • Buffer 2: 20 - 50%
  • Buffer 3: 0 - 20%
  • Buffer 4: 100%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 8.0 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 95% of the DNA fragments can be ligated and re-cut after 50-fold over-digestion with NheI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 5 units of NheI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Notes:
    • Low salt, high glycerol (>5%) concentrations, pH>8.0 or a large excess of enzyme may result in star activity.
    • NheI is inhibited by salt concentrations above 100mM.
    • Supercoiled plasmids may require up to 10-fold more NheI for complete digestion than linear DNAs (e.g. 10 units are required to cleave 1μg of pBR322 DNA).
    Compatible Ends: XmaJI, BcuI, Eco130I, XbaI
    Methylation Effects:
    Dam: Never overlaps - no effect
    Dcm: Never overlaps - no effect
    CpG: May overlap - cleavage impaired
    • 5'...GCTAGm5C G...3'
  • 3'...CGATC Gm5C...5'
  • (Cleavage is impaired)
    • 5'...m5C GCTAGm5C G...3'
  • 3'... Gm5CGATC Gm5C...5'
  • (Cleavage is blocked)
  • EcoKI: Never overlaps - no effect
  • EcoBI: Never overlaps - no effect
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 25 to 50% Water, >50% Glycerin
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nhe I
    Storage Buffer 10mM Tris HCl (pH 8 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 500 U
    Cut Site G.CTAGC
    Product Type NheI
    Form Liquid
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.