missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ NcoI, Fisher BioReagents™
Click to view available options
Quantity:
500 U
Unit Size:
Each
Description
5'...C^C A T G G...3'
3'...G G T A C^C...5'
Supplied with: 10X OPTIZYME Buffer 4
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 4: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 20 - 50%
- Buffer 2: 20 - 50%
- Buffer 3: 20 - 50%
- Buffer 4: 100%
- Buffer 5: 50 - 100%
- 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 25 to 50% Water, >50% Glycerin |
| pH | 7.4 |
| For Use With (Application) | >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nco I |
| Storage Buffer | 10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 500 U |
| Cut Site | C.CATGG |
| Product Type | NcoI |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction