missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ EcoRV (Eco32I), Fisher BioReagents™
Click to view available options
Quantity:
3000 U
Unit Size:
Each
Description
5'...G A T^A T C...3'
3'...C T A^T A G...5'
Supplied with: 10X OPTIZYME Buffer 5
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 0 - 20%
- Buffer 2: 50 - 100%
- Buffer 3: 50 - 100%
- Buffer 4: 20 - 50%
- Buffer 5: 100%
- 25mM Tris-HCl (pH 7.5 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- 5'...m5C GATATm5C G...3'
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRV |
| Storage Buffer | 25mM Tris HCl (pH 7.5 at 25°C), 200mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 3000 U |
| Cut Site | GAT.ATC |
| Product Type | EcoRV (Eco32I) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction