missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ EcoRI, Fisher BioReagents™

Product Code. p-4362819
Click to view available options
Quantity:
15,000 U
25,000 U
Concentration:
10 U/μL
50 U/μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11924842

Brand: Fisher BioReagents BP80035

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...G^A A T T C...3'
3'...C T T A A^G...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer EcoRI: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
  • Incubate at 37°C.
Supplied with: 10X OPTIZYME EcoRI Buffer
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: No reaction
  • Buffer 3: 100%
  • Buffer 4: No reaction
  • Buffer 5: 100%™
™star activity occurs at greater than 5-fold overdigestion (5 units X 1 hour)
Storage Buffer:
10mM potassium phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/ml BSA, 0.15% Triton X-100 and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with EcoRI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of EcoRI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt concentration, large excess of the enzyme, pH>8.0, or the replacement of mg2+ by Mn2+ may result in star activity.
Compatible Ends: XapI, MunI, TasI.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: never overlaps - no effect.
CpG: may overlap - cleavage impaired.
  • 5'...m5C GAATTm5C G...3'
  • 3'... Gm5CTTAA Gm5...5'
  • EcoKI: never overlaps - no effect.
  • EcoBI: may overlap - no effect.
    • 5'...TGm6AATTC(N)4TGCT...3'
  • 3'...AC TTAAG(N)4m6ACGA...5'
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Components 25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride
    Incubator Temperature 37°C
    pH 7.4
    For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI
    Storage Buffer 10mM Potassium Phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
    Content And Storage Keep container tightly closed
    Quantity 15,000 U
    Cut Site G.AATTC
    Product Type EcoRI
    Form Liquid
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.