missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ BglI, Fisher BioReagents™
Click to view available options
Quantity:
2000 U
Unit Size:
Each
Description
5'...G C C N N N N^N G G C...3'
3'...C G G N^N N N N C C G...5'
Supplied with: 10X OPTIZYME Buffer 3
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 3: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/ml BSA
- Incubate at 37°C
- Buffer 1: 0 - 20%
- Buffer 2: 50 - 100%
- Buffer 3: 100%
- Buffer 4: 0 - 20%
- Buffer 5: 100%
- 10mM Tris-HCl (pH 7.5 at 25°C), 300mM NaCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- 5'...GCm5CW GGNNGGC...3'
- 5'...GCm5C G(N)3m5C GGC...3'
- 5'...GCC(N)5GGm5C G...3'
- 5'...m5C GCC(N)5GGm5C G...3'
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 25 to 50% Water, >50% Glycerin |
| Incubator Temperature | 37°C |
| pH | 7.4 |
| For Use With (Application) | >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BglI |
| Storage Buffer | 10mM Tris HCl (pH 7.5 at 25°C), 300mM NaCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 2000 U |
| Cut Site | GCCNNNN.NGGC |
| Product Type | BglI |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction