missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPTIZYME™ BclI, Fisher BioReagents™
Click to view available options
Quantity:
3000 U
Unit Size:
Each
Description
5'...T^G A T C A...3'
3'...A C T A G^T...5'
Supplied with: 10X OPTIZYME Buffer 2
Conditions for 100% Activity:
- 1X OPTIZYME Buffer 2: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/ml BSA
- Incubate at 55°C
- Buffer 1: 20 - 50%
- Buffer 2: 100%
- Buffer 3: 20 - 50%
- Buffer 4: 100%™
- Buffer 5: 20 - 50%
Storage Buffer:
- 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol
- Incubation at 37°C results in 50% activity.
- Greater than 15-fold overdigestion with BclI may result in star activity.
- Assayed using lambda DNA (dam-)
- BclI is blocked by dam methylation. To avoid dam methylation, use a dam-, dcm- strain.
Methylation Effects:
Dam: Completely overlaps - blocked
- TGm6ATCA
- 5'...TGm6A TCA(N)5 TGCT...3'
Specifications
Specifications
| Concentration | 10 U/μL |
| Components | 25 to 50% Water, >50% Glycerin |
| Incubator Temperature | 55°C |
| pH | 7.4 |
| For Use With (Application) | >95% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with BclI |
| Storage Buffer | 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol |
| Content And Storage | Keep container tightly closed |
| Quantity | 3000 U |
| Cut Site | T.GATCA |
| Product Type | BcII |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction