missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ BamHI, Fisher BioReagents™

Product Code. 11974842
Change view
Click to view available options
Quantity:
12,500 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11974842 12,500 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11974842 Supplier Fisher BioReagents Supplier No. BP80055
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5'...G^G A T C C...3'
3'...C C T A G^G...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer BamHI: 10mM Tris-HCl (pH 8.0 at 37°C), 5mM MgCl2, 100mM KCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
  • Incubate at 37°C
Supplied with: 10X OPTIZYME BamHI Buffer
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 20 - 50%™
  • Buffer 2: 100%
  • Buffer 3: 20 - 50%
  • Buffer 4: 100%™
  • Buffer 5: 50 - 100%™
™star activity occurs at greater than 5-fold overdigestion (5 units × 1 hour)
Storage Buffer:
10mM Tris-HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with BamHI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of BamHI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.
Compatible Ends: BclI, BglII, Bsp143I, MboI, PsuI.
Methylation Effects:
Dam: completely overlaps - no effect.
Dcm: may overlap - no effect.
CpG: may overlap - no effect.
EcoKI: never overlaps - no effect.
EcoBI: never overlaps - no effect.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Components 25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
Incubator Temperature 37°C
pH 7.4
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BamHI
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
Content And Storage Keep container tightly closed
Quantity 12,500 U
Cut Site G.GATCC
Product Type BamHI
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.